Stampa…

(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer ≥96.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Dichloro(pentamethylcyclopentadienyl)rhodium (III) dimer, [Rh(Cp-)Cl2]2, Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer, Dichloro(pentamethylcyclopentadienyl)rhodium(III) Dimer

P1788-200MG P1788-1G
TCIAP1788-200MGEA 113 EUR
TCIAP1788-200MG TCIAP1788-1G
(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer ≥96.0% (by titrimetric analysis)
(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer
Formula: C₂₀H₃₀Cl₄Rh₂
Peso molecolare: 618,07 g/mol
MDL Number: MFCD00061552
CAS Number: 12354-85-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89d4a594d6d9ce165a442e7fafc4aeb7e272e939&vid=89d4a594d6d9ce165a442e7fafc4aeb7e272e939&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Argentmetric Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR