Stampa…

2-Phenyl-2-(2-pyridyl)acetamide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Phenyl-2-(pyridin-2-yl)acetamide

P1741-25G P1741-5G
TCIAP1741-25GEA 186 EUR
TCIAP1741-25G TCIAP1741-5G
2-Phenyl-2-(2-pyridyl)acetamide ≥98.0% (by HPLC, titration analysis)
2-Phenyl-2-(2-pyridyl)acetamide
Formula: C₁₃H₁₂N₂O
Peso molecolare: 212,25 g/mol
MDL Number: MFCD00138126
CAS Number: 7251-52-7
EINECS: 230-663-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 134.0 to 138.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR