D-Phenylalanine benzyl ester-p-toluenesulphonate ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
Synonyms:
H-D-Phe-OBzl.Tos, H-D-Phe-OBzl.TosOH, D-Phenylalanine benzyl ester tosylate
Formula:
C₂₃H₂₅NO₅S Peso molecolare: 427.52 g/mol |
MDL Number:
MFCD00066129 CAS Number: 28607-46-7 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | +5.0 to +7.0 deg (C=2, methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...