Stampa…

Phenyl trifluoromethyl sulphide ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Phenyl(trifluoromethyl)sulphane, alpha,alpha,alpha-Trifluorothioanisole, (Trifluoromethylthio)benzene

P1693-25G P1693-5G
TCIAP1693-25GEA 169 EUR
TCIAP1693-25G TCIAP1693-5G
Phenyl trifluoromethyl sulphide ≥97.0% (by GC)
Phenyl trifluoromethyl sulphide
Formula: C₇H₅F₃S
Peso molecolare: 178,17 g/mol
Punto di ebollizione: 142°C
Flash Pt: 37°C
MDL Number: MFCD00040839
CAS Number: 456-56-4
EINECS: 207-270-0
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a78eff143c981089e37260d2659a4d1623347dca&vid=a78eff143c981089e37260d2659a4d1623347dca&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR