Stampa…

Phenethyl isocyanate ≥98.0% (by GC)

Fornitore: TCI
Pericolo

Synonyms: Isocyanic Acid Phenethyl Ester, (2-Isocyanatoethyl)benzene, Isocyanic Acid 2-Phenylethyl Ester, 2-Phenylethyl Isocyanate

P1677-25G P1677-5G
TCIAP1677-25GEA 71 EUR
TCIAP1677-25G TCIAP1677-5G
Phenethyl isocyanate ≥98.0% (by GC)
Phenethyl isocyanate
Formula: C₉H₉NO
Punto di ebollizione: 210°C
MDL Number: MFCD00037044
CAS Number: 1943-82-4
EINECS: 413-080-0
UN: 2206
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR