Phenyl-3,4,6-tri-O-acetyl-2-deoxy-1-thio-2-(2,2,2-trichloroethoxyformamido)-β-D-galactopyranoside ≥98.0% (by HPLC)
Fornitore: TCI
Formula:
C₂₁H₂₄Cl₃NO₉S Temperatura di stoccaggio: Refrigerata |
CAS Number:
278784-83-1 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 115.0 to 119.0 deg-C |
Specific rotation [a]20/D | -4.0 to -6.0 deg (C=1、CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...