Stampa…

4-(trans-4-Propylcyclohexyl)phenol ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4-(Trans-4-propylcyclohexyl)phenol, 1-Hydroxy-4-(trans-4-propylcyclohexyl)benzene

P1616-25G P1616-100G
TCIAP1616-25GEA 76 EUR
TCIAP1616-25G TCIAP1616-100G
4-(trans-4-Propylcyclohexyl)phenol ≥98.0% (by GC)
4-(trans-4-Propylcyclohexyl)phenol
Formula: C₁₅H₂₂O
Peso molecolare: 218,34 g/mol
Punto di fusione: 138°C
MDL Number: MFCD00673751
CAS Number: 81936-33-6
EINECS: 440-740-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 136.0 to 142.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR