Stampa…

4-Phenylthiomorpholine-1,1-dioxide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

P1603-5G P1603-25G
TCIAP1603-5GEA 140 EUR
TCIAP1603-5G TCIAP1603-25G
4-Phenylthiomorpholine-1,1-dioxide ≥98.0% (by GC)
4-Phenylthiomorpholine-1,1-dioxide
Formula: C₁₀H₁₃NO₂S
Peso molecolare: 211,28 g/mol
CAS Number: 17688-68-5
EINECS: 241-675-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b859fe6cc6ed229d1d43afcdc274e04acefa6bb&vid=9b859fe6cc6ed229d1d43afcdc274e04acefa6bb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Purple powder to crystal
Purity (GC) min. 98.0 %
Melting point 123.0 to 127.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR