Stampa…

2-(2-Pyridyl)benzimidazole ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: AI-4-88

P1599-5G P1599-1G
TCIAP1599-5GEA 350 EUR
TCIAP1599-5G TCIAP1599-1G
2-(2-Pyridyl)benzimidazole ≥98.0% (by HPLC, titration analysis)
2-(2-Pyridyl)benzimidazole
Formula: C₁₂H₉N₃
Peso molecolare: 195,23 g/mol
MDL Number: MFCD00005586
CAS Number: 1137-68-4
EINECS: 214-508-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79538e794d1ff84829ccecde391d428be5ff3607&vid=79538e794d1ff84829ccecde391d428be5ff3607&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 223.0 to 227.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR