Stampa…

Trimethoxy[3-(phenylamino)propyl]silane ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: N-[3-(Trimethoxysilyl)propyl]aniline

P1458-25G P1458-250G
TCIAP1458-25GEA 24 EUR
TCIAP1458-25G TCIAP1458-250G
Trimethoxy[3-(phenylamino)propyl]silane ≥98.0% (by titrimetric analysis)
Trimethoxy[3-(phenylamino)propyl]silane
Formula: C₁₂H₂₁NO₃Si
Peso molecolare: 255,39 g/mol
Punto di ebollizione: 310°C
Flash Pt: 165°C
MDL Number: MFCD00053673
CAS Number: 3068-76-6
EINECS: 221-328-2
UN: 3267
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR