Stampa…

Potassium tris(3,5-dimethylpyrazol-1-yl)borohydride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Potassium Hydrotris(3,5-dimethylpyrazol-1-yl)borate, Potassium hydrotris(3,5-dimethylpyrazol-1-yl)borate

P1441-25G P1441-5G
TCIAP1441-25GEA 380 EUR
TCIAP1441-25G TCIAP1441-5G
Potassium tris(3,5-dimethylpyrazol-1-yl)borohydride ≥98.0% (by titrimetric analysis)
Potassium tris(3,5-dimethylpyrazol-1-yl)borohydride
Formula: C₁₅H₂₂BKN₆
Peso molecolare: 336,29 g/mol
MDL Number: MFCD00040342
CAS Number: 17567-17-8
UN: 2813
ADR: 4.3,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR