Stampa…

Potassium tris(1-pyrazolyl)borohydride ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Potassium Hydrotris(1-pyrazolyl)borate

P1440-1G P1440-5G
TCIAP1440-1GEA 30 EUR
TCIAP1440-1G TCIAP1440-5G
Potassium tris(1-pyrazolyl)borohydride ≥97.0% (by titrimetric analysis)
Potassium tris(1-pyrazolyl)borohydride
Formula: C₉H₁₀BKN₆
Peso molecolare: 252,13 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00040346
CAS Number: 18583-60-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR