Stampa…
Poly(styrene) on resin, cross-linked with 1% DVB (100-200 mesh)
Fornitore: TCI EUROPE
P1432-100G P1432-25G
TCIAP1432-100GEA 172 EUR
TCIAP1432-100G TCIAP1432-25G
Poly(styrene) on resin, cross-linked with 1% DVB (100-200 mesh)
Poly(styrene)
Formula: C₈H₈
Peso molecolare: 125000 - 250000 g/mol
MDL Number: MFCD00084450
CAS Number: 9003-53-6
Visualizza i codici
Appearance White to Almost white powder to crystal
Grain size 100 to 200 mesh