Stampa…

Poly[4-(diacetoxyiodo)styrene]

Fornitore: TCI EUROPE

Synonyms: Polystyrene Iodosoacetate, PSDIB

P1415-1G P1415-5G
TCIAP1415-1GEA 132 EUR
TCIAP1415-1G TCIAP1415-5G
Poly[4-(diacetoxyiodo)styrene]
Poly[4-(diacetoxyiodo)styrene]
MDL Number: MFCD06798811
CAS Number: 36290-94-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR