Stampa…

L-Prolinamide ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: H-Pro-NH₂, (S)-2-Pyrrolidinecarboxamide, H-Pro-NH2

P1382-25G P1382-5G P1382-1G
TCIAP1382-25GEA 110 EUR
TCIAP1382-25G TCIAP1382-5G TCIAP1382-1G
L-Prolinamide ≥97.0% (by titrimetric analysis)
L-Prolinamide
Formula: C₅H₁₀N₂O
Peso molecolare: 114,15 g/mol
Punto di fusione: 99°C
MDL Number: MFCD00005253
CAS Number: 7531-52-4
EINECS: 231-397-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24ecccf359e003aac337c4b02c564a05461a05d3&vid=24ecccf359e003aac337c4b02c564a05461a05d3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 98.0 to 102.0 deg-C
Specific rotation [a]20/D -103.0 to -109.0 deg (C=2, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR