Stampa…

ɑ-Pinene oxide ≥95.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2,3-Epoxypinane, alpha-Pinene oxide

P1362-25ML P1362-250ML
TCIAP1362-25MLEA 29 EUR
TCIAP1362-25ML TCIAP1362-250ML
ɑ-Pinene oxide ≥95.0% (by GC)
alpha-Pinene oxide
Formula: C₁₀H₁₆O
Peso molecolare: 152,24 g/mol
Flash Pt: 66°C
MDL Number: MFCD00066955
CAS Number: 1686-14-2
EINECS: 216-869-6
UN: 2810
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR