1,4-Benzenediacrylic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
1,4-Phenylenediacrylic acid, 1,4-Benzenediacrylic acid
Formula:
C₁₂H₁₀O₄ Peso molecolare: 218.21 g/mol Punto di fusione: 300°C |
MDL Number:
MFCD00002698 CAS Number: 16323-43-6 EINECS: 240-399-0 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...