Diallyl dicarbonate ≥95.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Diallyl Pyrocarbonate, Pyrocarbonic Acid Diallyl Ester
P1277-1G
P1277-5G
TCIAP1277-1GEA
53
EUR
TCIAP1277-1G
TCIAP1277-5G
Diallyl dicarbonate ≥95.0% (by GC)
Diallyl dicarbonate
Formula:
C₈H₁₀O₅ Peso molecolare: 186,16 g/mol Flash Pt: 93°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00077721 CAS Number: 115491-93-5 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...