Stampa…

Methyl-2-pyridylacetate ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Pyridineacetic Acid Methyl Ester, Methyl 2-Pyridineacetate, Methyl 2-(pyridin-2-yl)acetate, 2-Pyridylacetic Acid Methyl Ester

P1268-25G P1268-5G
TCIAP1268-25GEA 202 EUR
TCIAP1268-25G TCIAP1268-5G
Methyl-2-pyridylacetate ≥97.0% (by GC)
Methyl-2-pyridylacetate
Formula: C₈H₉NO₂
Peso molecolare: 151,17 g/mol
Punto di ebollizione: 137°C
MDL Number: MFCD00006358
CAS Number: 1658-42-0
EINECS: 216-759-8

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR