Stampa…

Diethyl-2,6-pyridinedicarboxylate ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Dipicolinic Acid Diethyl Ester, 2,6-Pyridinedicarboxylic Acid Diethyl Ester, Diethyl pyridine-2,6-dicarboxylate, Diethyl Dipicolinate

P1240-25G P1240-5G
TCIAP1240-25GEA 116 EUR
TCIAP1240-25G TCIAP1240-5G
Diethyl-2,6-pyridinedicarboxylate ≥98.0% (by GC, titration analysis)
Diethyl-2,6-pyridinedicarboxylate
Formula: C₁₁H₁₃NO₄
Peso molecolare: 223,23 g/mol
Punto di ebollizione: 188°C
MDL Number: MFCD00033792
CAS Number: 15658-60-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=869c7f004ca7ea8758862135577471a01edb6f28&vid=869c7f004ca7ea8758862135577471a01edb6f28&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 43.0 to 46.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR