Stampa…

3-(3-Pyridyl)acrylic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Pyridineacrylic Acid, 3-(Pyridin-3-yl)acrylic acid

P1157-25G P1157-5G
TCIAP1157-25GEA 152 EUR
TCIAP1157-25G TCIAP1157-5G
3-(3-Pyridyl)acrylic acid ≥98.0% (by titrimetric analysis)
3-(3-Pyridyl)acrylic acid
Formula: C₈H₇NO₂
Peso molecolare: 149,15 g/mol
MDL Number: MFCD00006410
CAS Number: 1126-74-5
EINECS: 214-424-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 234.0 to 238.0 deg-C
Solubility in Pyridine (50%) almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR