Stampa…

Monobutyl phthalate ≥95.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 2-(Butoxycarbonyl)benzoic acid, Butyl Hydrogen Phthalate, Phthalic Acid Monobutyl Ester

P1132-25G P1132-5G
TCIAP1132-25GEA 119 EUR
TCIAP1132-25G TCIAP1132-5G
Monobutyl phthalate ≥95.0% (by GC, titration analysis)
Monobutyl phthalate
Formula: C₁₂H₁₄O₄
MDL Number: MFCD00043498
CAS Number: 131-70-4
EINECS: 205-036-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 72.0 to 76.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR