Stampa…

1,3-Diaminopropane-N,N'-diacetic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI

Synonyms: 1,3-Propylenediamine-N,N'-diacetic Acid

P1115-25G P1115-5G
TCIAP1115-25GEA 106 EUR
TCIAP1115-25G TCIAP1115-5G
1,3-Diaminopropane-N,N'-diacetic acid ≥98.0% (by titrimetric analysis)
1,3-Diaminopropane-N,N'-diacetic acid
Formula: C₇H₁₄N₂O₄
Peso molecolare: 190.20 g/mol
MDL Number: MFCD00142534
CAS Number: 112041-05-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR