1-(2-Pyrimidyl)piperazine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-(1-Piperazinyl)pyrimidine, 2-(Piperazin-1-yl)pyrimidine
P1110-25G
P1110-5G
TCIAP1110-25GEA
66
EUR
TCIAP1110-25G
TCIAP1110-5G
1-(2-Pyrimidyl)piperazine ≥98.0% (by GC)
1-(2-Pyrimidyl)piperazine
Formula:
C₈H₁₂N₄ Peso molecolare: 164,21 g/mol Punto di ebollizione: 120°C |
MDL Number:
MFCD00040742 CAS Number: 20980-22-7 EINECS: 244-135-5 |
Risultati del test di specificazione
Appearance | White or Colorless to Light orange to Yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...