Stampa…

4-Pentyloxyphthalonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,2-Dicyano-4-pentyloxybenzene, 4-Amyloxyphthalonitrile, 4-(Pentyloxy)phthalonitrile

P1071-1G P1071-5G
TCIAP1071-1GEA 67 EUR
TCIAP1071-1G TCIAP1071-5G
4-Pentyloxyphthalonitrile ≥98.0% (by GC)
4-Pentyloxyphthalonitrile
Formula: C₁₃H₁₄N₂O
Peso molecolare: 214,27 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00059103
CAS Number: 106943-83-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 35.0 to 37.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR