Stampa…

4-Propoxyphthalonitrile ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,2-Dicyano-4-propoxybenzene

P1070-1G P1070-5G
TCIAP1070-1GEA 86 EUR
TCIAP1070-1G TCIAP1070-5G
4-Propoxyphthalonitrile ≥95.0% (by GC)
4-Propoxyphthalonitrile
Formula: C₁₁H₁₀N₂O
Peso molecolare: 186,21 g/mol
MDL Number: MFCD00059102
CAS Number: 106144-18-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR