Pamoic acid disodium salt monohydrate ≥98.0% (by HPLC)
Fornitore: TCI
Synonyms:
4,4'-Methylenebis(3-hydroxy-2-naphthoic Acid Sodium Salt), Embonic Acid Disodium Salt
Formula:
C₂₃H₁₆O₆ Peso molecolare: 432.34 g/mol |
MDL Number:
MFCD00036183 CAS Number: 6640-22-8 EINECS: 229-653-1 |
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 95.0 % (calcd.on anh.substance) |
Water | 3.0 to 5.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...