Stampa…

Methyl-4,4-dimethyl-3-oxovalerate ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Pivaloylacetic Acid Methyl Ester, 4,4-Dimethyl-3-oxovaleric Acid Methyl Ester, Methyl 4,4-dimethyl-3-oxopentanoate, Methyl Pivaloylacetate

P1031-25ML P1031-500ML P1031-100ML
TCIAP1031-25MLEA 68 EUR
TCIAP1031-25ML TCIAP1031-500ML TCIAP1031-100ML
Methyl-4,4-dimethyl-3-oxovalerate ≥95.0% (by GC)
Methyl-4,4-dimethyl-3-oxovalerate
Formula: C₈H₁₄O₃
Peso molecolare: 158,20 g/mol
Punto di ebollizione: 84°C
Flash Pt: 77°C
MDL Number: MFCD00008847
CAS Number: 55107-14-7
EINECS: 259-481-2

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR