Copper(II) phthalocyanine ≥93.0% (by titrimetric analysis)
Fornitore: TCI
Formula:
C₃₂H₁₆CuN₈ Punto di fusione: 480°C |
MDL Number:
MFCD00010719 CAS Number: 147-14-8 EINECS: 205-685-1 Merck Index: 14,02520 |
|
Risultati del test di specificazione
Appearance | Dark red to Dark purple to Dark blue powder to crystal |
Purity (Chelometric Titration) | min. 93.0 % |
Lambda max. | 789.0 to 791.0 nm (H2SO4) |
Molar extinction coefficient | min. 220,000 (H2SO4, 789.0 to 791.0 nm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...