Stampa…

Copper(II) phthalocyanine ≥90.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: 29H,31H-Phthalocyaninato-N29,N30,N31,N32 copper, Pigment Blue 15

P1005-250G P1005-25G
TCIAP1005-250GEA 134 EUR
TCIAP1005-250G TCIAP1005-25G
Copper(II) phthalocyanine ≥90.0% (by titrimetric analysis)
Copper(II) phthalocyanine
Formula: C₃₂H₁₆CuN₈
Peso molecolare: 576,08 g/mol
Punto di fusione: 480°C
MDL Number: MFCD00010719
CAS Number: 147-14-8
EINECS: 205-685-1
Merck Index: 14,02520

Visualizza i codici

Risultati del test di specificazione

Appearance Blue to Dark blue powder to crystal
Purity (Chelometric Titration) min. 90.0 %
Lambda max. 790.0 to 792.0 nm (H2SO4)
Molar extinction coefficient min. 210,000 (H2SO4, 790.0 to 792.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR