3,4,9,10-Perylenetetracarboxylic dianhydride ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Anthra[2,1,9-def:6,5,10-d'e'f']diisochromene-1,3,8,10-tetraone, Pigment Red 224, PTCDA
P0972-25G
P0972-500G
P0972-100G
TCIAP0972-25GEA
39
EUR
TCIAP0972-25G
TCIAP0972-500G
TCIAP0972-100G
3,4,9,10-Perylenetetracarboxylic dianhydride ≥98.0% (by titrimetric analysis)
3,4,9,10-Perylenetetracarboxylic dianhydride
Formula:
C₂₄H₈O₆ Peso molecolare: 392,32 g/mol |
MDL Number:
MFCD00006916 CAS Number: 128-69-8 EINECS: 204-905-3 |
Risultati del test di specificazione
Appearance | Orange to Brown to Dark red powder to crystal |
Purity (Neutralization back titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...