Stampa…

Prazosin hydrochloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-(2-furanylcarbonyl)piperazine Hydrochloride, (4-(4-Amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl)(furan-2-yl)methanone hydrochloride, Prazosin HCl

P0938-1G P0938-100MG
TCIAP0938-1GEA 177 EUR
TCIAP0938-1G TCIAP0938-100MG
Prazosin hydrochloride ≥98.0% (by titrimetric analysis)
Prazosin hydrochloride
Formula: C₁₉H₂₁N₅O₄
Peso molecolare: 419,87 g/mol
Punto di fusione: 285°C
MDL Number: MFCD00058177
CAS Number: 19237-84-4
EINECS: 242-903-4
Merck Index: 14,07717

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR