Stampa…

5-Dimethylamino-2-(2-pyridylazo)phenol ≥98.0% (by HPLC) for determination of zinc in serum

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-(2-Pyridylazo)-5-dimethylaminophenol

P0910-1G P0910-100MG
TCIAP0910-1GEA 327 EUR
TCIAP0910-1G TCIAP0910-100MG
5-Dimethylamino-2-(2-pyridylazo)phenol ≥98.0% (by HPLC) for determination of zinc in serum
5-Dimethylamino-2-(2-pyridylazo)phenol
Formula: C₁₃H₁₄N₄O
Peso molecolare: 242,28 g/mol
MDL Number: MFCD00059778
CAS Number: 50783-80-7

Visualizza i codici

Risultati del test di specificazione

Appearance Red to Dark red to Dark purple powder to crystal
Purity (HPLC) min. 98.0 area%
Lambda max. 569.0 to 573.0 nm (EtOH (50vol%))
Molar extinction coefficient (Co complex) min. 75000 (EtOH (50vol%), 569.0 to 573.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR