Pentaerythritol-tetra(3-mercaptopropionate) ≥90.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₇H₂₈O₈S₄ |
MDL Number:
MFCD00022104 CAS Number: 7575-23-7 EINECS: 231-472-8 UN: 3082 ADR: 9,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (Iodometric back titration) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...