Stampa…

Methyl pentadecanoate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Pentadecanoic Acid Methyl Ester

P0869-25ML P0869-5ML
TCIAP0869-25MLEA 274 EUR
TCIAP0869-25ML TCIAP0869-5ML
Methyl pentadecanoate ≥98.0% (by GC)
Methyl pentadecanoate
Formula: C₁₆H₃₂O₂
Peso molecolare: 256,43 g/mol
Punto di ebollizione: 155°C
MDL Number: MFCD00008989
CAS Number: 7132-64-1
EINECS: 230-430-6

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR