Polyethylene glycol monocetyl ether n=∼23
Fornitore: TCI
P0841-25G
P0841-500G
TCIAP0841-25GEA
18
EUR
TCIAP0841-25G
TCIAP0841-500G
Polyethylene glycol monocetyl ether n=∼23
Polyethylene glycol monocetyl ether
Formula:
HO(CH₂CH₂O)nC₁₆H₃₃ |
MDL Number:
MFCD00081419 CAS Number: 9004-95-9 EINECS: 500-014-1 |
|
Risultati del test di specificazione
Appearance | White to Almost white wax |
The average value of n | 21.0 to 25.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...