Dipropyl-2,5-pyridinedicarboxylate ≥98.0% (by GC)
Fornitore: TCI
P0835-25G
P0835-5G
TCIAP0835-25GEA
314
EUR
TCIAP0835-25G
TCIAP0835-5G
Dipropyl-2,5-pyridinedicarboxylate ≥98.0% (by GC)
Dipropyl-2,5-pyridinedicarboxylate
Formula:
C₁₃H₁₇NO₄ Punto di ebollizione: 175°C |
MDL Number:
MFCD00055315 CAS Number: 136-45-8 EINECS: 205-245-9 |
Risultati del test di specificazione
Appearance | Colorless to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...