Stampa…

Iron(II) phthalocyanine ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: Phthalocyanine Iron(II), Iron phthalocyanine

P0774-25G P0774-5G
TCIAP0774-25GEA 87 EUR
TCIAP0774-25G TCIAP0774-5G
Iron(II) phthalocyanine ≥97.0% (by titrimetric analysis)
Iron(II) phthalocyanine
Formula: C₃₂H₁₆FeN₈
Peso molecolare: 568,38 g/mol
MDL Number: MFCD00015953
CAS Number: 132-16-1
EINECS: 205-047-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3332f292f5ab155f2e11d529b2e6b6bbe46c004c&vid=3332f292f5ab155f2e11d529b2e6b6bbe46c004c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Dark purple powder to crystal
Purity (Chelometric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR