Stampa…

4-(3-Phenylpropyl)piperidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

P0760-25G P0760-5G
TCIAP0760-25GEA 169 EUR
TCIAP0760-25G TCIAP0760-5G
4-(3-Phenylpropyl)piperidine ≥98.0% (by GC, titration analysis)
4-(3-Phenylpropyl)piperidine
Formula: C₁₄H₂₁N
Peso molecolare: 203,33 g/mol
MDL Number: MFCD00023152
CAS Number: 18495-82-4
EINECS: 242-381-8

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR