Polyethylene glycol stearylamine n=∼10
Fornitore: TCI EUROPE
P0724-500G
P0724-25G
TCIAP0724-500GEA
46
EUR
TCIAP0724-500G
TCIAP0724-25G
Polyethylene glycol stearylamine n=∼10
Polyethylene glycol stearylamine
Risultati del test di specificazione
Appearance | Red to Dark red to Brown clear liquid |
Purity (Neutralization titration) | min. 90.0 % |
The average value of n | 8.0 to 12.0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...