Stampa…

5β-Pregnane-3ɑ,20ɑ-diol

Fornitore: TCI

P0678-1G P0678-100MG
TCIAP0678-1GEA 286 EUR
TCIAP0678-1G TCIAP0678-100MG
5β-Pregnane-3ɑ,20ɑ-diol
5beta-Pregnane-3alpha,20alpha-diol
Formula: C₂₁H₃₆O₂
MDL Number: MFCD00064141
CAS Number: 80-92-2
EINECS: 201-313-7
Merck Index: 14,07727

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +25.0 to +28.0 deg (C=0.5, EtOH)
Solubility in EtOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR