Stampa…

2-(Pyridin-3-yl)acetic acid hydrochloride ≥99.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Pyridyl-3-acetic acid hydrochloride, 3-Pyridylacetic acid hydrochloride

P0639-10G P0639-25G
TCIAP0639-10GEA 56 EUR
TCIAP0639-10G TCIAP0639-25G
2-(Pyridin-3-yl)acetic acid hydrochloride ≥99.0% (by titrimetric analysis)
2-(Pyridin-3-yl)acetic acid hydrochloride
Formula: C₇H₈ClNO₂
Peso molecolare: 173,60 g/mol
MDL Number: MFCD00012819
CAS Number: 6419-36-9
EINECS: 229-148-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Purity (Argentmetric Titration) min. 99.0 %
Melting point 157.0 to 162.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR