2-Phenoxypropionyl chloride ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₉H₉ClO₂ Punto di ebollizione: 106°C |
MDL Number:
MFCD00018810 CAS Number: 122-35-0 EINECS: 204-536-8 UN: 3265 ADR: 8,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...