Stampa…

Pentamethoxy Red

Fornitore: TCI EUROPE

Synonyms: Bis(2,4-dimethoxyphenyl)(2-methoxyphenyl)methanol, 2,2',2'',4,4'-Pentamethoxytriphenylmethanol

P0631-1G P0631-25G
TCIAP0631-1GEA 93 EUR
TCIAP0631-1G TCIAP0631-25G
Pentamethoxy Red
Pentamethoxy Red
Formula: C₂₄H₂₆O₆
Peso molecolare: 410,47 g/mol
MDL Number: MFCD00070616
CAS Number: 1755-51-7
EINECS: 217-146-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90b936450eeaad00f8e172757b267208c66cf6e9&vid=90b936450eeaad00f8e172757b267208c66cf6e9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 145.0 to 148.0 deg-C
Transition interval (pH) (red purple)1.2 to 3.8 (colorless)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR