Stampa…

N-Phthaloyl-L-glutamic anhydride ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: (S)-2-(2,6-Dioxotetrahydro-2H-pyran-3-yl)isoindoline-1,3-dione

P0620-1G P0620-5G
TCIAP0620-1GEA 34 EUR
TCIAP0620-1G TCIAP0620-5G
N-Phthaloyl-L-glutamic anhydride ≥98.0% (by GC, titration analysis)
N-Phthaloyl-L-glutamic anhydride
Formula: C₁₃H₉NO₅
Peso molecolare: 259,22 g/mol
MDL Number: MFCD00067324
CAS Number: 25830-77-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Morpholine Method) min. 98.0 %
Specific rotation [a]20/D -41.0 to -44.0 deg (C=3, Dioxane)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR