Pontacyl carmine 2B ≥93.0% (by HPLC)
Fornitore: TCI EUROPE
P0595-25G
P0595-500G
TCIAP0595-25GEA
27
EUR
TCIAP0595-25G
TCIAP0595-500G
Pontacyl carmine 2B ≥93.0% (by HPLC)
Pontacyl carmine 2B
Formula:
C₁₉H₁₇N₃O₉S₂ |
MDL Number:
MFCD00059131 CAS Number: 6625-46-3 EINECS: 229-584-7 |
Risultati del test di specificazione
Appearance | Red to Dark red to Brown powder to crystal |
Purity (HPLC) | min. 93.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...