Stampa…

Phenyl propionate ≥97.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Propionic Acid Phenyl Ester

P0509-25G P0509-100G
TCIAP0509-25GEA 56 EUR
TCIAP0509-25G TCIAP0509-100G
Phenyl propionate ≥97.0% (by GC)
Phenyl propionate
Formula: C₉H₁₀O₂
Peso molecolare: 150,18 g/mol
Punto di ebollizione: 211°C
Flash Pt: 86°C
MDL Number: MFCD00053765
CAS Number: 637-27-4
EINECS: 211-282-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d797b4208c744ee8943fc4e2f19786e414da3edb&vid=d797b4208c744ee8943fc4e2f19786e414da3edb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR