Stampa…

Piperazine dihydrochloride monohydrate ≥98.0% (by total nitrogen basis)

Fornitore: TCI EUROPE
Pericolo

P0450-25G P0450-500G
TCIAP0450-25GEA 21 EUR
TCIAP0450-25G TCIAP0450-500G
Piperazine dihydrochloride monohydrate ≥98.0% (by total nitrogen basis)
Piperazine dihydrochloride monohydrate
Formula: C₄H₁₀N₂·2HCl·H₂O
Peso molecolare: 177,07 g/mol
Punto di fusione: 320°C
MDL Number: MFCD00012754
CAS Number: 142-64-3
EINECS: 205-551-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water 9.0 to 12.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR