Pinacyanol iodide ≥90.0% (by HPLC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Iodide, Quinaldine Blue, 1,1'-Diethyl-2,2'-carbocyanine Iodide
P0436-1G
P0436-5G
TCIAP0436-1GEA
57
EUR
TCIAP0436-1G
TCIAP0436-5G
Pinacyanol iodide ≥90.0% (by HPLC)
Pinacyanol iodide
Formula:
C₂₅H₂₅IN₂ Peso molecolare: 480,39 g/mol |
MDL Number:
MFCD00011975 CAS Number: 605-91-4 EINECS: 210-099-4 UN: 2811 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Green to Dark green powder to crystal |
Purity (HPLC) | min. 90.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...