Stampa…

L-Phenylephrine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (R)-(-)-1-(3-Hydroxyphenyl)-2-(methylamino)ethanol, (R)-(-)-3-[1-Hydroxy-2-(methylamino)ethyl]phenol, (R)-(-)-3-Hydroxy-alpha-(methylaminomethyl)benzyl Alcohol, (R)-(-)-Phenylephrine

P0395-25G P0395-5G
TCIAP0395-25GEA 193 EUR
TCIAP0395-25G TCIAP0395-5G
L-Phenylephrine ≥98.0% (by HPLC, titration analysis)
L-Phenylephrine
Formula: C₉H₁₃NO₂
Peso molecolare: 167,21 g/mol
MDL Number: MFCD00044749
CAS Number: 59-42-7
EINECS: 200-424-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 172.0 to 176.0 deg-C
Specific rotation [a]20/D -53.0 to -58.0 deg (C=5, 1mol/L HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR